2,4-Dimethoxybenzylamin

Lieferant: COMBI-BLOCKS

Synonyme: 1-(2,4-Dimethoxyphenyl)methanamine, (2,4-Dimethoxyphenyl)methanamine

OR-0696-25G
COBBOR-0696-25GEA 79.3 EUR
COBBOR-0696-25G
2,4-Dimethoxybenzylamin
2,4-Dimethoxybenzylamin
Formel: C₉H₁₃NO₂
Molecular Weight: 167,21 g/mol
MDL: MFCD00052393
CAS-Nummer: 20781-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR