2-Chlor-4,6-dimethylpyrimidin

Lieferant: COMBI-BLOCKS

OR-0821-25G
COBBOR-0821-25GEA 96.1 EUR
COBBOR-0821-25G
2-Chlor-4,6-dimethylpyrimidin
2-Chloro-4,6-dimethylpyrimidine
Formel: C₆H₇ClN₂
Molecular Weight: 142,59 g/mol
MDL: MFCD00023199
CAS-Nummer: 4472-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR