3-Iodbenzonitril

Lieferant: COMBI-BLOCKS

OR-0932-25G
COBBOR-0932-25GEA 122 EUR
COBBOR-0932-25G
3-Iodbenzonitril
3-Iodbenzonitril
Formel: C₇H₄IN
Molecular Weight: 229,02 g/mol
MDL: MFCD00079762
CAS-Nummer: 69113-59-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR