N-Boc-L-prolinol
Lieferant: COMBI-BLOCKS
Synonyme:
N-(tert-Butoxycarbonyl)-L-prolinol, 1-Boc-2-(S)-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol, N-Boc-L-Prolinol
OR-1188-25G
COBBOR-1188-25GEA
45.6
EUR
COBBOR-1188-25G
N-Boc-L-prolinol
N-Boc-L-prolinol
Formel:
C₁₀H₁₉NO₃ Molecular Weight: 201,27 g/mol |
MDL:
MFCD00066232 CAS-Nummer: 69610-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...