Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate

OR-1285-100G
COBBOR-1285-100GEA 0 EUR
COBBOR-1285-100G
Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat
Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat
Formel: C₈H₉ClN₂O₂S
MDL: MFCD00006085
CAS-Nummer: 5909-24-0
EINECS: 227-619-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR