7-Chlorisatin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 7-Chlorindolin-2,3-dion, 7-Chlor-2,3-dioxoindolin, 7-Chlor-1H-indol-2,3-dion, 7-Chlor-2,3-indolindion

OR-1448-5G
COBBOR-1448-5GEA 0 EUR
COBBOR-1448-5G
7-Chlorisatin
7-Chlorisatin
Formel: C₈H₄ClNO₂
MDL: MFCD00022796
CAS-Nummer: 7477-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR