N-Boc-D-prolinol

Lieferant: COMBI-BLOCKS

Synonyme: (R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, N-tert-Butoxycarbonyl-D-prolinol, N-t-Boc-D-prolinol, Boc-D-Prolinol

OR-1478-25G
COBBOR-1478-25GEA 90.7 EUR
COBBOR-1478-25G
N-Boc-D-prolinol
N-Boc-D-prolinol
Formel: C₁₀H₁₉NO₃
Molecular Weight: 201,27 g/mol
MDL: MFCD00040580
CAS-Nummer: 83435-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR