3,5-Difluor-2-pyridincarbonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyan-3,5-difluorpyridin, 3,5-Difluor-2-pyridincarbonsäurenitril

OR-1548-25G
COBBOR-1548-25GEA 462 EUR
COBBOR-1548-25G
3,5-Difluor-2-pyridincarbonitril
3,5-Difluor-2-pyridincarbonitril
Formel: C₆H₂F₂N₂
Molecular Weight: 140,09 g/mol
MDL: MFCD03788759
CAS-Nummer: 298709-29-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR