(R)-(-)-1-Methoxy-2-propanol

Lieferant: COMBI-BLOCKS

Synonyme: (R)-(-)-Propylenglycolmonomethylether

OR-1566-25G
COBBOR-1566-25GEA 108 EUR
COBBOR-1566-25G
(R)-(-)-1-Methoxy-2-propanol
(R)-(-)-1-Methoxy-2-propanol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632587
CAS-Nummer: 4984-22-9
UN: 3092
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR