4,6-Dichlor-5-methoxypyrimidin

Lieferant: COMBI-BLOCKS

OR-1627-5G
COBBOR-1627-5GEA 51.3 EUR
COBBOR-1627-5G
4,6-Dichlor-5-methoxypyrimidin
4,6-Dichloro-5-methoxypyrimidine
Formel: C₅H₄Cl₂N₂O
Molecular Weight: 179,01 g/mol
MDL: MFCD00087680
CAS-Nummer: 5018-38-2
EINECS: 225-699-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR