1-Methyl-4-piperidinmethanol

Lieferant: COMBI-BLOCKS

OR-1747-25G
COBBOR-1747-25GEA 109 EUR
COBBOR-1747-25G
1-Methyl-4-piperidinmethanol
1-Methyl-4-piperidinmethanol
Formel: C₇H₁₅NO
MDL: MFCD03411903
CAS-Nummer: 20691-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR