m-β-Hydroxyethylsulfonylanilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-[(3-Aminophenyl)sulphonyl]ethanol

OR-1846-5G
COBBOR-1846-5GEA 357 EUR
COBBOR-1846-5G
m-β-Hydroxyethylsulfonylanilin
m-β-Hydroxyethylsulfonylanilin
Formel: C₈H₁₁NO₃S
MDL: MFCD00044070
CAS-Nummer: 5246-57-1
EINECS: 226-046-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR