2-Amino-4-methoxypyridin

Lieferant: COMBI-BLOCKS

OR-2142-5G
COBBOR-2142-5GEA 97.2 EUR
COBBOR-2142-5G
2-Amino-4-methoxypyridin
2-Amino-4-methoxypyridine
Formel: C₆H₈N₂O
Molecular Weight: 124,14 g/mol
MDL: MFCD07437849
CAS-Nummer: 10201-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR