Methyl-2-(brommethyl)benzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-bromomethylbenzoate

OR-2562-5G
COBBOR-2562-5GEA 131 EUR
COBBOR-2562-5G
Methyl-2-(brommethyl)benzoat
Methyl-2-(brommethyl)benzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,0706 g/mol
MDL: MFCD03425900
CAS-Nummer: 2417-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR