2-Methoxy-4-methyl-5-nitropyridin

Lieferant: COMBI-BLOCKS

OR-5581-5G
COBBOR-5581-5GEA 61.3 EUR
COBBOR-5581-5G
2-Methoxy-4-methyl-5-nitropyridin
2-Methoxy-4-methyl-5-nitropyridine
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
MDL: MFCD03095075
CAS-Nummer: 6635-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR