3-(Brommethyl)thiophen

Lieferant: COMBI-BLOCKS

OR-5999-1G
COBBOR-5999-1GEA 0 EUR
COBBOR-5999-1G
3-(Brommethyl)thiophen
3-(Bromomethyl)thiophene
Formel: C₅H₅BrS
Molecular Weight: 177,06 g/mol
MDL: MFCD03840497
CAS-Nummer: 34846-44-1
EINECS: 252-247-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR