3-Amino-2-chlor-4-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Amino-2-chloro-4-methylpyridine, 3-Amino-2-chloro-4-picoline, 2-Chloro-4-methylpyridin-3-amine

OS-1015-25G
COBBOS-1015-25GEA 50.8 EUR
COBBOS-1015-25G
3-Amino-2-chlor-4-methylpyridin
3-Amino-2-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142,59 g/mol
MDL: MFCD00673152
CAS-Nummer: 133627-45-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR