3,6-Dichlor-4-methylpyridazin
Lieferant: COMBI-BLOCKS
|
Achtung
|
OS-1032-100G
COBBOS-1032-100GEA
218
EUR
COBBOS-1032-100G
3,6-Dichlor-4-methylpyridazin
3,6-Dichloro-4-methylpyridazine
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,01 g/mol Siedepunkt: 130…135 °C (1 mmHg) Schmelzpunkt: 87 °C |
MDL:
MFCD00006465 CAS-Nummer: 19064-64-3 EINECS: 242-794-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...