1-Brom-3,5-di-tert-butylbenzol

Lieferant: COMBI-BLOCKS

OS-1034-100G
COBBOS-1034-100GEA 0 EUR
COBBOS-1034-100G
1-Brom-3,5-di-tert-butylbenzol
1-Brom-3,5-di-tert-butylbenzol
Formel: C₁₄H₂₁Br
MDL: MFCD00796945
CAS-Nummer: 22385-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR