4-Amino-3-nitropyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

OS-1062-25G
COBBOS-1062-25GEA 113 EUR
COBBOS-1062-25G
4-Amino-3-nitropyridin
4-Amino-3-nitropyridin
Formel: C₅H₅N₃O₂
Molecular Weight: 139,11 g/mol
MDL: MFCD00160304
CAS-Nummer: 1681-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR