5-Iod-o-toluylsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-lodo-2-methylbenzoic acid, 5-Iodo-2-methylbenzoic acid

OS-1080-5G
COBBOS-1080-5GEA 23.2 EUR
COBBOS-1080-5G
5-Iod-o-toluylsäure
5-Iod-o-toluylsäure
Formel: C₈H₇IO₂
Molecular Weight: 262,0400 g/mol
MDL: MFCD01570258
CAS-Nummer: 54811-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR