Methyl-2-brom-3-methylbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-bromo-3-methylbenzoate

OS-1092-1G
COBBOS-1092-1GEA 26.2 EUR
COBBOS-1092-1G
Methyl-2-brom-3-methylbenzoat
Methyl-2-brom-3-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229.07 g/mol
MDL: MFCD00144762
CAS-Nummer: 131001-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR