1,3-Dichlor-2-nitrobenzol

Lieferant: COMBI-BLOCKS

OS-1154-100G
COBBOS-1154-100GEA 279 EUR
COBBOS-1154-100G
1,3-Dichlor-2-nitrobenzol
1,3-Dichlor-2-nitrobenzol
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
MDL: MFCD00456795
CAS-Nummer: 601-88-7
EINECS: 210-009-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR