4-Brom-2,6-difluorbenzonitril

Lieferant: COMBI-BLOCKS

OS-2093-25G
COBBOS-2093-25GEA 0 EUR
COBBOS-2093-25G
4-Brom-2,6-difluorbenzonitril
4-Brom-2,6-difluorbenzonitril
Formel: C₇H₂BrF₂N
Molecular Weight: 218 g/mol
MDL: MFCD01310981
CAS-Nummer: 123843-67-4
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR