2-Brom-α,α,α,6-tetrafluortoluol

Lieferant: COMBI-BLOCKS

OS-2101-25G OS-2101-100G
COBBOS-2101-25GEA 0 EUR
COBBOS-2101-25G COBBOS-2101-100G
2-Brom-α,α,α,6-tetrafluortoluol
2-Brom-α,α,α,6-tetrafluortoluol
Formel: C₇H₃BrF₄
MDL: MFCD01631569
CAS-Nummer: 261951-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR