2-Brom-3-fluor-6-methylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
2-Brom-3-fluor-6-picolin
OS-2126-25G
COBBOS-2126-25GEA
484
EUR
COBBOS-2126-25G
2-Brom-3-fluor-6-methylpyridin
2-Bromo-3-fluoro-6-methylpyridine
Formel:
C₆H₅BrFN Molecular Weight: 190,01 g/mol |
MDL:
MFCD03095061 CAS-Nummer: 374633-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...