3-Amino-2-brom-5-fluorpyridin

Lieferant: COMBI-BLOCKS

OS-2186-5G
COBBOS-2186-5GEA 0 EUR
COBBOS-2186-5G
3-Amino-2-brom-5-fluorpyridin
3-Amino-2-brom-5-fluorpyridin
Formel: C₅H₄BrFN₂
MDL: MFCD05662413
CAS-Nummer: 884495-03-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR