2-Brom-4-(trifluormethyl)benzoesäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-4-(trifluormethyl)benzoesäure

OS-2246-5G
COBBOS-2246-5GEA 0 EUR
COBBOS-2246-5G
2-Brom-4-(trifluormethyl)benzoesäure
2-Brom-4-(trifluormethyl)benzoesäure
Formel: C₈H₄BrF₃O₂
MDL: MFCD07368837
CAS-Nummer: 328-89-2
EINECS: 206-339-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR