2-Brom-5-iodtoluol

Lieferant: COMBI-BLOCKS
Achtung

OS-2362-100G
COBBOS-2362-100GEA 445 EUR
COBBOS-2362-100G
2-Brom-5-iodtoluol
2-Brom-5-iodtoluol
Formel: C₇H₆BrI
Molecular Weight: 296,93 g/mol
MDL: MFCD00070749
CAS-Nummer: 202865-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR