2-Amino-5-brom-3-iodpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Brom-3-iodpyridin-2-amin, 5-Brom-3-iod-2-pyridylamin
OS-2412-5G
COBBOS-2412-5GEA
86.2
EUR
COBBOS-2412-5G
2-Amino-5-brom-3-iodpyridin
2-Amino-5-brom-3-iodpyridin
Formel:
C₅H₄BrIN₂ Molecular Weight: 298,91 g/mol |
MDL:
MFCD06659000 CAS-Nummer: 381233-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...