2-Amino-6-methoxy-3-nitropyridin
Lieferant: COMBI-BLOCKS
Synonyme:
6-Methoxy-3-nitropyridin-2-amine
OS-7409-5G
OS-7409-25G
OS-7409-100G
COBBOS-7409-5GEA
26.2
EUR
COBBOS-7409-5G
COBBOS-7409-25G
COBBOS-7409-100G
2-Amino-6-methoxy-3-nitropyridin
2-Amino-6-methoxy-3-nitropyridine
Formel:
C₆H₇N₃O₃ Molecular Weight: 169.14 g/mol |
MDL:
MFCD03206481 CAS-Nummer: 73896-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...