2,6-Dichlor-5-fluornicotinonitril

Lieferant: COMBI-BLOCKS

OS-7416-25G OS-7416-100G OS-7416-5G
COBBOS-7416-25GEA 0 EUR
COBBOS-7416-25G COBBOS-7416-100G COBBOS-7416-5G
2,6-Dichlor-5-fluornicotinonitril
2,6-Dichlor-5-fluornicotinonitril
Formel: C₆HCl₂FN₂
MDL: MFCD01863289
CAS-Nummer: 82671-02-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR