3-(3-Indolyl)propionsäure
Lieferant: COMBI-BLOCKS
OS-7428-25G
OS-7428-100G
COBBOS-7428-25GEA
0
EUR
COBBOS-7428-25G
COBBOS-7428-100G
3-(3-Indolyl)propionsäure
3-(3-Indolyl)propionsäure
Formel:
C₁₁H₁₁NO₂ Schmelzpunkt: 129…132 °C Dichte: 1,356 g/cm³ (20 °C) |
MDL:
MFCD00005660 CAS-Nummer: 830-96-6 EINECS: 212-600-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...