2-Brom-3-methylpyridin

Lieferant: COMBI-BLOCKS
Achtung

OS-7437-5G OS-7437-25G OS-7437-100G
COBBOS-7437-5GEA 0 EUR
COBBOS-7437-5G COBBOS-7437-25G COBBOS-7437-100G
2-Brom-3-methylpyridin
2-Brom-3-methylpyridin
Formel: C₆H₆BrN
MDL: MFCD00239380
CAS-Nummer: 3430-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR