2-Brom-6-methylpyridin

Lieferant: COMBI-BLOCKS
Achtung

OS-7438-25G OS-7438-100G
COBBOS-7438-25GEA 0 EUR
COBBOS-7438-25G COBBOS-7438-100G
2-Brom-6-methylpyridin
2-Brom-6-methylpyridin
Formel: C₆H₆BrN
MDL: MFCD00040743
CAS-Nummer: 5315-25-3
EINECS: 226-173-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR