1-Brom-2-fluor-4-nitrobenzol

Lieferant: COMBI-BLOCKS

OS-7449-5G OS-7449-25G OS-7449-100G
COBBOS-7449-5GEA 0 EUR
COBBOS-7449-5G COBBOS-7449-25G COBBOS-7449-100G
1-Brom-2-fluor-4-nitrobenzol
1-Bromo-2-fluoro-4-nitrobenzene
Formel: C₆H₃BrFNO₂
MDL: MFCD05865088
CAS-Nummer: 185331-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR