3-Chlor-2-nitroanilin
Lieferant: COMBI-BLOCKS
|
Gefahr
|
OS-7452-5G
OS-7452-25G
OS-7452-1G
COBBOS-7452-5GEA
0
EUR
COBBOS-7452-5G
COBBOS-7452-25G
COBBOS-7452-1G
3-Chlor-2-nitroanilin
3-Chlor-2-nitroanilin
Formel:
C₆H₅ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01317803 CAS-Nummer: 59483-54-4 EINECS: 261-782-9 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...