(4-Bromphenyl)essigsäure
Lieferant: COMBI-BLOCKS
OS-7455-500G
OS-7455-25G
OS-7455-100G
COBBOS-7455-500GEA
0
EUR
COBBOS-7455-500G
COBBOS-7455-25G
COBBOS-7455-100G
(4-Bromphenyl)essigsäure
(4-Bromphenyl)essigsäure
Formel:
(C₆H₄Br)CH₂COOH Siedepunkt: 326 °C (1013 hPa) Schmelzpunkt: 94…98 °C |
MDL:
MFCD00004342 CAS-Nummer: 1878-68-8 EINECS: 217-523-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...