4-Brom-2-methylpyridin
Lieferant: COMBI-BLOCKS
OS-7471-25G
OS-7471-100G
OS-7471-5G
COBBOS-7471-25GEA
140
EUR
COBBOS-7471-25G
COBBOS-7471-100G
COBBOS-7471-5G
4-Brom-2-methylpyridin
4-Brom-2-methylpyridin
Formel:
C₆H₆BrN Molecular Weight: 172.02 g/mol |
MDL:
MFCD03788196 CAS-Nummer: 22282-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...