2-Brom-5-fluoranilin

Lieferant: COMBI-BLOCKS
Achtung

OS-7479-25G OS-7479-100G
COBBOS-7479-25GEA 0 EUR
COBBOS-7479-25G COBBOS-7479-100G
2-Brom-5-fluoranilin
2-Brom-5-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
MDL: MFCD00070750
CAS-Nummer: 1003-99-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR