3-Brom-4-fluoranilin

Lieferant: COMBI-BLOCKS

OS-7480-25G OS-7480-5G OS-7480-100G
COBBOS-7480-25GEA 50.8 EUR
COBBOS-7480-25G COBBOS-7480-5G COBBOS-7480-100G
3-Brom-4-fluoranilin
3-Brom-4-fluoranilin
Formel: C₆H₅BrFN
MDL: MFCD03788559
CAS-Nummer: 656-64-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR