Methyl-2-amino-3-thiophencarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-aminothiophene-3-carboxylate

OS-7483-5G OS-7483-25G OS-7483-100G
COBBOS-7483-5GEA 26 EUR
COBBOS-7483-5G COBBOS-7483-25G COBBOS-7483-100G
Methyl-2-amino-3-thiophencarboxylat
Methyl-2-amino-3-thiophencarboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157.19 g/mol
MDL: MFCD00159547
CAS-Nummer: 4651-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR