2-Aminopyrazin
Lieferant: COMBI-BLOCKS
Synonyme:
Aminopyrazin, Pyrazin-2-amin
OS-7492-25G
OS-7492-100G
COBBOS-7492-25GEA
0
EUR
COBBOS-7492-25G
COBBOS-7492-100G
2-Aminopyrazin
2-Aminopyrazine
Formel:
C₄H₅N₃ Siedepunkt: 203 °C (1013 hPa) Schmelzpunkt: 117…120 °C |
MDL:
MFCD00006137 CAS-Nummer: 5049-61-6 EINECS: 225-748-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...