2-Chlor-4-nitropyridin

Lieferant: COMBI-BLOCKS

OS-7495-5G OS-7495-25G OS-7495-1G
COBBOS-7495-5GEA 0 EUR
COBBOS-7495-5G COBBOS-7495-25G COBBOS-7495-1G
2-Chlor-4-nitropyridin
2-Chloro-4-nitropyridine
Formel: C₅H₃ClN₂O₂
Schmelzpunkt: 53,5…54 °C
MDL: MFCD00661454
CAS-Nummer: 23056-36-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR