2-(Trifluormethoxy)benzoesäure

Lieferant: COMBI-BLOCKS
Achtung

OS-7502-100G OS-7502-25G OS-7502-5G
COBBOS-7502-100GEA 312 EUR
COBBOS-7502-100G COBBOS-7502-25G COBBOS-7502-5G
2-(Trifluormethoxy)benzoesäure
2-(Trifluormethoxy)benzoesäure
Formel: C₈H₅F₃O₃
Molecular Weight: 206.12 g/mol
MDL: MFCD00052325
CAS-Nummer: 1979-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR