2-Brom-6-fluoranilin

Lieferant: COMBI-BLOCKS
Achtung

OS-7514-5G OS-7514-25G OS-7514-100G
COBBOS-7514-5GEA 43.4 EUR
COBBOS-7514-5G COBBOS-7514-25G COBBOS-7514-100G
2-Brom-6-fluoranilin
2-Brom-6-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
MDL: MFCD01310982
CAS-Nummer: 65896-11-9
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR