Chlor(1,5-cyclooctadien)iridium(I) dimer

Lieferant: COMBI-BLOCKS

Synonyme: [Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer

OS-7525-1G
COBBOS-7525-1GEA 445 EUR
COBBOS-7525-1G
Chlor(1,5-cyclooctadien)iridium(I) dimer
Chlor(1,5-cyclooctadien)iridium(I) dimer
Formel: C₁₆H₂₄Cl₂Ir₂
Molecular Weight: 671,70 g/mol
MDL: MFCD00012414
CAS-Nummer: 12112-67-3
EINECS: 235-170-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR