2-Pyrimidincarbonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyanopyrimidine

OS-7535-5G OS-7535-25G OS-7535-100G
COBBOS-7535-5GEA 0 EUR
COBBOS-7535-5G COBBOS-7535-25G COBBOS-7535-100G
2-Pyrimidincarbonitril
2-Pyrimidincarbonitril
Formel: C₅H₃N₃
Molecular Weight: 105.1 g/mol
MDL: MFCD00160513
CAS-Nummer: 14080-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR