2-Pyrimidincarbonitril
Lieferant: COMBI-BLOCKS
Synonyme:
2-Cyanopyrimidine
OS-7535-5G
OS-7535-25G
OS-7535-100G
COBBOS-7535-5GEA
0
EUR
COBBOS-7535-5G
COBBOS-7535-25G
COBBOS-7535-100G
2-Pyrimidincarbonitril
2-Pyrimidincarbonitril
Formel:
C₅H₃N₃ Molecular Weight: 105.1 g/mol |
MDL:
MFCD00160513 CAS-Nummer: 14080-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...