2,6-Di-tert-butyl-4-methylpyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2,6-Di-Tert-butyl-4-methylpyridine

OS-7542-25G OS-7542-5G OS-7542-1G
COBBOS-7542-25GEA 399 EUR
COBBOS-7542-25G COBBOS-7542-5G COBBOS-7542-1G
2,6-Di-tert-butyl-4-methylpyridin
2,6-Di-tert-butyl-4-methylpyridin
Formel: C₁₄H₂₃N
Molecular Weight: 205.34 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00006305
CAS-Nummer: 38222-83-2
EINECS: 253-834-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR